Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Human: 86%; Mouse: 100%; Rabbit: 86%; Rat: 86%
Gonadal dysgenesis in 9p-deleted individuals might be due to combined hemizygosity of DMRT1 and DMRT2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dmrt2; Doublesex- and mab-3-related transcription factor 2; Doublesex-like 2 protein; doublesex-like 2 protein (DSXL-2); Terra; terra homolog; terra like protein antibody
Gene Aliases: Dmrt2; Terra
UniProt ID: (Mouse) Q8BG36
Entrez Gene ID: (Mouse) 226049
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support