Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is predicted to react with bovine based on sequence homology.
DMRTA2 may be involved in sexual development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DMRT-like family A2; DMRT5; doublesex and mab-3 related transcription factor like family A2; Doublesex- and mab-3-related transcription factor 5; Doublesex- and mab-3-related transcription factor A2
Gene Aliases: DMRT5; DMRTA2
UniProt ID: (Human) Q96SC8, (Mouse) A2A9A2
Entrez Gene ID: (Human) 63950, (Mouse) 242620, (Zebrafish) 474350
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support