Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This gene encodes a transcription factor that contains a cyclin D-binding domain, three central Myb-like repeats, and two flanking acidic transactivation domains at the N- and C-termini. The encoded protein is induced by the oncogenic Ras signaling pathway and functions as a tumor suppressor by activating the transcription of ARF and thus the ARF-p53 pathway to arrest cell growth or induce apoptosis. It also activates the transcription of aminopeptidase N and may play a role in hematopoietic cell differentiation. The transcriptional activity of this protein is regulated by binding of D-cyclins. This gene is hemizygously deleted in approximately 40% of human non-small-cell lung cancer and is a potential prognostic and gene-therapy target for non-small-cell lung cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cyclin D binding myb-like transcription factor 1; cyclin D binding myb-like transcription factor 1D-interacting myb-like protein; cyclin D-binding Myb-like protein; Cyclin-D-binding Myb-like transcription factor 1; Cyclin-D-interacting Myb-like protein 1; D-interacting myb-like protein; FLJ41265; hDMP1; hDMTF1; mDmp1
Gene Aliases: Dimp; DMP1; DMTF; DMTF1; hDMP1; MRUL
UniProt ID: (Human) Q9Y222, (Rat) Q66HG1, (Mouse) Q8CE22
Entrez Gene ID: (Human) 9988, (Rat) 114485, (Mouse) 23857
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support