Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CDPRHKDCLL REFRKLCAMV ADNPSYNTKT QIIQDFLRKG SAGDGFHGDV YLTVKLLLPG VIKTVYNLND KQIVKLFSRI FNCNPDDMAR DLEQGDVSET IRVFFEQSKS FPPAAKSLLT IQEVDEFLLR LSKLTKE
Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA Ligase III gene product migrates as a 100 kDa (a-form) protein in somatic tissues, is ubiquitously expressed and complexes with XRCC-1 and a 97 kDa (beta form) protein that is found in testis. DNA Ligase III is involved in rejoining DNA single-strand breaks and in the base excision repair pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support