Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to DNA2.
Immunogen sequence: MEQLNELEL LMEKSFWEEA ELPAELFQKK VVASFPRTVL STGMDNRYLV LAVNTVQNKE GNCEKRLVIT ASQSLENKEL CILRNDWCSV PVEPGDIIHL EGDCTSDTWI IDKDFGYLIL YPDMLISGTS IASSIRCMRR AVLSETFRSS DPATRQMLIG TVLHEVFQKA INNSFAPEKL QELAFQTIQE IRHLKEMYRL NLSQDEIKQE VEDYLPSFCK WAGDFMHKNT STDFPQMQLS LPSDNSKDNS TCNIEVVKPM DIEESIWSPR FGLKGKIDVT VGVKIHRGYK TKYKIMPLEL KTGKESNSIE HRSQVVLYTL LSQERRADPE AGLLLYLKTG QMYPVPANHL D (1-350 aa encoded by BC111740)
DNA2 is a conserved helicase/nuclease involved in themaintenance of mitochondrial and nuclear DNA stability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA replication ATP-dependent helicase-like homolog; DNA replication ATP-dependent helicase/nuclease DNA2; DNA replication helicase 2 homolog; DNA2-like helicase; hDNA2
Gene Aliases: DNA2; DNA2L; E130315B21Rik; hDNA2; KIAA0083
UniProt ID: (Human) P51530, (Mouse) Q6ZQJ5
Entrez Gene ID: (Human) 1763, (Mouse) 327762
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support