Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 92%; Dog: 93%; Guinea Pig: 85%; Horse: 86%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 85%; Rat: 86%
DNA2 is a conserved helicase/nuclease involved in themaintenance of mitochondrial and nuclear DNA stability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA replication ATP-dependent helicase-like homolog; DNA replication ATP-dependent helicase/nuclease DNA2; DNA replication helicase 2 homolog; DNA2-like helicase; hDNA2
Gene Aliases: DNA2; DNA2L; hDNA2; KIAA0083
UniProt ID: (Human) P51530
Entrez Gene ID: (Human) 1763
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support