Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to DNAH6.
Immunogen sequence: MTYTEQASK RHYCMRLTCF IRLNDYLIEN TMHILTVNAV NSLLNHLTDK LKRTPSADVI QKWITEEKPE VPDKKGTLMV EKQEEDESLI PMFLTELMLT VQSLLFEPSL EDFLDGILGA VNHCQNTVLS VPNLVPDSYF DAFTSPYINN KLEGKTCGTG PSLAAVFEDD KNFHTIISQI KETIQAAFES ARIYAATFEK FQIFFKENES LDLQALKLQE PDINFFSEQL EKYHKQHKDA VALRPTRNVG LLLIDTRLLR EKLIPSPLRC LEVLNFMLPR QSKKKVDAII FEAQDAEYKL EFVPTTTTEY VHSLLFLDEI QERIESLEDE GNIVTQMYKL MEQYQVPTPP EDFAVFATMK PSIVAVRNAI DKSVGDRESS IKQFCVHLGS DLEELNNEVN EVKLQAQVS (1-408 aa encoded by BC104884 )
DNAH6 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axonemal beta dynein heavy chain 6; axonemal dynein heavy chain; Ciliary dynein heavy chain 6; Dynein axonemal heavy chain 6; dynein heavy chain 6, axonemal; dynein heavy chain-like 1; dynein, axon, heavy chain 6; dynein, axonemal, heavy polypeptide 6
Gene Aliases: 9830168K20; A730004I20Rik; axo a; DNAH6; DNAHC6; DNHL1; HL-2; HL2; KIAA1697; mdhc6; mKIAA1697
UniProt ID: (Human) Q9C0G6
Entrez Gene ID: (Human) 1768, (Mouse) 330355, (Rat) 117250
Molecular Function:
microtubule binding motor protein
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support