Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DNAH6 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axonemal beta dynein heavy chain 6; Ciliary dynein heavy chain 6; Dynein axonemal heavy chain 6; dynein heavy chain-like 1; dynein, axonemal, heavy polypeptide 6
Gene Aliases: DNAH6; DNAHC6; DNHL1; HL-2; HL2; KIAA1697
UniProt ID: (Human) Q9C0G6
Entrez Gene ID: (Human) 1768
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support