Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axonemal beta dynein heavy chain 8; Ciliary dynein heavy chain 8; Dynein axonemal heavy chain 8; dynein, axonemal, heavy chain 8; dynein, axonemal, heavy polypeptide 8
Gene Aliases: ATPase; DNAH8; hdhc9
UniProt ID: (Human) Q96JB1
Entrez Gene ID: (Human) 1769
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support