Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: DFYSLLGVSK TASSREIRQA FKKLALKLHP DKNPNNPNAH GDFLKINRAY
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%
This endoplasmic reticulum co-chaperone may play a role in protein folding and translocation across the endoplasmic reticulum membrane. DNAJC10 may act as a co-chaperone for HSPA5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily C, member 10; DnaJ homolog subfamily C member 10; Endoplasmic reticulum DNA J domain-containing protein 5; ER-resident protein ERdj5; J-domain-containing protein disulfide isomerase-like protein; Macrothioredoxin; MTHr; protein disulfide isomerase family A, member 19
Gene Aliases: DNAJC10; ERDJ5; JPDI; MTHr; PDIA19; UNQ495/PRO1012
UniProt ID: (Human) Q8IXB1
Entrez Gene ID: (Human) 54431
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support