Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: QVRAQTAELF AKMTADKLIG PKVRITLMKF LPSVFMDAMR DNPEAAVHIF EGTHENPELI WNDNSRDKVS TTVREMM
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
DNAJC13, also known as receptor-mediated endocytosis 8 (RME8), is the human homolog to a DnaJ domain-containing protein originally identified in a screen for endocytic defects in C. elegans. It is thought to be a co-chaperone of Hsc70 which regulates protein conformation at membrane sites and plays a role in intracellular trafficking, co-localizing with markers of the endosomal system. Recent experiments have indicated that the DNAJC13 protein is involved in membrane trafficking through early endosomes but not through degradative organelles. DNAJC13 has been also been shown to regulate the intracellular trafficking of the epidermal growth factor receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily C, member 13; DnaJ domain-containing protein RME-8; DnaJ homolog subfamily C member 13; Required for receptor-mediated endocytosis 8; RME-8
Gene Aliases: DNAJC13; KIAA0678; PARK21; RME8
UniProt ID: (Human) O75165
Entrez Gene ID: (Human) 23317
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support