Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Peptide Sequence: RYVLQAMKHM EPQVKQVFQS LPKSAFSGGY YRGGFEPKMT KREAALILGV SPTANKGKIR DAHRRIMLLN HPDKGGSPYI AAKINEAKDL LEGQAKK
The protein encoded by this gene is thought to be part of a complex involved in the ATP-dependent transport of transit peptide-containing proteins from the inner cell membrane to the mitochondrial matrix. Defects in this gene are a cause of 3-methylglutaconic aciduria type 5 (MGA5), also known as dilated cardiomyopathy with ataxia (DCMA). Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 1, 2, 6, 10, 14 and 19. [provided by RefSeq, Jan 2012]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily C, member 19; DnaJ homolog subfamily C member 19; DnaJ-like protein subfamily C member 19; homolog of yeast TIM14; Mitochondrial import inner membrane translocase subunit TIM14
Gene Aliases: DNAJC19; PAM18; TIM14; TIMM14
UniProt ID: (Human) Q96DA6
Entrez Gene ID: (Human) 131118
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support