Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
GTPase which can activate the MEK/ERK pathway and induce cell transformation when overexpressed. May act as a nuclear scaffold for MAPK1, probably by association with MAPK1 nuclear export signal leading to enhanced ERK1/ERK2 signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APAH1; DnaJ (Hsp40) homolog, subfamily C, member 27; DnaJ homolog subfamily C member 27; Rab and DnaJ domain-containing protein; Ras-associated protein Rap1
Gene Aliases: DNAJC27; RABJS; RBJ
UniProt ID: (Human) Q9NZQ0
Entrez Gene ID: (Human) 51277
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support