Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAATEPELLD DQEAKREAET FKEQGNAYYA KKDYNEAYNY YTKAIDMCPK NASYYGNRAA TLMMLGRFRE ALGDAQQSVR LDDSFVRGHL REGKCHLSLG NAMAACRSFQ RALELDHKNA QAQQEFKNAN AVMEYEKIAE TDFEKRDFRK VVFCMDRALE FAPACHRFKI LKAECLAMLG RYPEAQSVAS DILRMDSTNA DALYVRGLCL YYEDCIEKAV QFFVQALRMA PDHEKACIAC RNAKALKAKK EDGNKAFKEG NYKLAYELYT EALGIDPNNI KTNAKLYCNR GTVNSKLRKL DDAIEDCTNA VKLDDTYIKA YLRRAQCYMD TEQYEEAVRD YEKVYQTEKT KEHKQLLKNA QLELKKSKRK DYYKILGVDK NASEDEIKKA YRKRALMHHP DRHSGASAEV QKEEEKKFKE VGEAFTILSD PKKKTRYDSG QDLDEEGMNM GDFDPNNIFK AFFGGPGGFS FEASGPGNFF FQFG
DNAJC7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily C, member 7; DnaJ homolog subfamily C member 7; tetratricopeptide repeat domain 2; Tetratricopeptide repeat protein 2; TPR repeat protein 2
Gene Aliases: DJ11; DJC7; DNAJC7; TPR2; TTC2
UniProt ID: (Human) Q99615
Entrez Gene ID: (Human) 7266
Molecular Function: chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support