Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: RRGWLTINNI SLMKGGSKEY WFVLTAESLS WYKDEEEKEK KYMLPLDNLK IRDVEKGFMS NKHVFAIFNT EQRNVYKDLR QIELACDSQE DVDSWKASFL RAGVYPEKDQ AENEDGAQEN TFSMDPQLER QVETIRNLVD SYVAIINKSI RDLMPKTIMH LMINNTKAFI HHELLAYLYS SADQSSLMEE SADQAQRRDD MLRMYHALKE ALNIIGDIST STVSTPVPPP VDDTWLQSAS SHSPTPQRRP VSSIHPPGRP PAVRGPTPGP PLIPVPVGAA ASFSAPPIPS RPGPQSVFAN SDLFPAPPQI PSRPVRIPPG IPPGVPSRRP PAAPSRPTII RPAEPSLLD
Dynamins represent one of the subfamilies of GTP-binding proteins. These proteins share considerable sequence similarity over the N-terminal portion of the molecule, which contains the GTPase domain. Dynamins are associated with microtubules. They have been implicated in cell processes such as endocytosis and cell motility, and in alterations of the membrane that accompany certain activities such as bone resorption by osteoclasts. Dynamins bind many proteins that bind actin and other cytoskeletal proteins. Dynamins can also self-assemble, a process that stimulates GTPase activity. Five alternatively spliced transcripts encoding different proteins have been described. Additional alternatively spliced transcripts may exist, but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dynamin 2; Dynamin II; Dynamin UDNM; Dynamin-2
Gene Aliases: b2b2159Clo; CMT2M; CMTDI1; CMTDIB; DI-CMTB; DNM2; DYIIAAB; DYN2; DYNII; LCCS5; Udnm
UniProt ID: (Human) P50570, (Rat) P39052, (Mouse) P39054
Entrez Gene ID: (Rabbit) 100348451, (Human) 1785, (Pig) 100622538, (Rat) 25751, (Mouse) 13430
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support