Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: MKGDTRHLNG EEDAGGREDS ILVNGACSDQ SSDSPPILEA IRTPEIRGRR SSSRLSKREV SSLLSYTQDL TGDGDGEDGD GSDTPVMPKL FRETRTRSES PAVRTRNNNS VSSRERHRPS PRSTRGRQGR NHVDESPVEF PATRSLRRRA TASAGTPWPS PPSSYLTIDL TDDTEDTHGT PQSSSTPYAR LAQDSQQGGM ESPQVEADSG DGDSSEYQDG KEFGIGDLVW GKIKGFSWWP AMVVSWKATS KRQAMSGMRW VQWFGDGKFS EVSADKLVAL GLFSQHFNLA TFNKLVSYRK AMYHALEKAR VRAGKTFPSS PGDSLEDQLK PMLEWAHGGF KPTGIEGLKP NNTQPVVNKS KVRRAGSRKL ESRKYENKTR RRTADDSATS DYCPAPKRLK TNCYNNGKDR
CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. This gene encodes a DNA methyltransferase which is thought to function in de novo methylation, rather than maintenance methylation. The protein localizes primarily to the nucleus and its expression is developmentally regulated. Mutations in this gene cause the immunodeficiency-centromeric instability-facial anomalies (ICF) syndrome. Six alternatively spliced transcript variants have been described. The full length sequences of variants 4 and 5 have not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA (cytosine-5)-methyltransferase 3B; DNA (cytosine-5-)-methyltransferase 3 beta; DNA methyltransferase HsaIIIB; DNA methyltransferase MmuIIIB; DNA MTase HsaIIIB; DNA MTase MmuIIIB; Dnmt3; Dnmt3b; m.MmuIIIB; MGC1090
Gene Aliases: DNMT3B; ICF; ICF1; M.HsaIIIB; MmuIIIB
UniProt ID: (Human) Q9UBC3, (Mouse) O88509
Entrez Gene ID: (Human) 1789, (Mouse) 13436
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support