Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: NQVNEQSAPL PVPVPVPVPS YGGEEPVRKE SKTPPPYSVY ERTLRRPVPL PHSLSIPVTS EPPALPPKPL AARSSHLENG ARRTDPGPRP RPLPRKVSQL
SRC2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dedicator of cytokinesis protein 4
Gene Aliases: DOCK4; KIAA0716
UniProt ID: (Human) Q8N1I0
Entrez Gene ID: (Human) 9732
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support