Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a guanine nucleotide exchange factor (GEF) that plays a role in axon formation and neuronal polarization. The encoded protein displays GEF activity toward RAC1 and RAC3 Rho small GTPases but not toward CDC42. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dedicator of cytokinesis protein 7; protein moonlight
Gene Aliases: 3110056M06Rik; DOCK7; EIEE23; Gm430; KIAA1771; m; ZIR2
UniProt ID: (Human) Q96N67
Entrez Gene ID: (Human) 85440, (Mouse) 67299, (Rat) 313388
Molecular Function: guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support