Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MACNQSAVYL QHCFATQRAL VSKFPELLFE EETEQCADLC LRLLRHCSSS IGTIRSHASA SLYLLMRQNF EIGNNFARVK MQVTMSLSSL VGTSQNFNEE FLRRSLKTIL TYAEEDLELR ETTFPDQVQD LVFNLHMILS DTVKMKEHQE DPEMLIDLMY RIAKGYQTSP DLRLTWLQNM AGKHSERSNH AEAAQCLVHS AALVAEYLSM LEDRKYLPVG CVTFQNISSN VLEESAVSDD VVSPDEEGIC SGKYFTESGL VGLLEQAAAS FSMAGMYEAV NEVYKVLIPI HEANRDAKKL STIHGKLQEA FSKIVHQDGK RMFGTYFRVG FYGTKFGDLD EQEFVYKEPA ITKLAEISHR LEGFYGERFG EDVVEVIKDS NPVDKCKLDP NKAYIQITYV EPYFDTYEMK DRITYFDKNY NLRRFMYCTP FTLDGRAHGE LHEQFKRKTI LTTSHAFPYI KTRVNVTHKE EIILTPIEVA IEDMQKKTQE LAFATHQDPA DPKMLQMVLQ GSVGTTVNQG PLEVAQVFLS EIPSDPKLFR HHNKLRLCFK DFTKRCEDAL RKNKSLIGPD QKEYQRELER NYHRLKEALQ PLINRKIPQL YKAVLPVTCH RDSFSRMSLR KMDL
The protein encoded by this gene is a guanine nucleotide exchange factor (GEF) that plays a role in axon formation and neuronal polarization. The encoded protein displays GEF activity toward RAC1 and RAC3 Rho small GTPases but not toward CDC42. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dedicator of cytokinesis protein 7
Gene Aliases: DOCK7; EIEE23; KIAA1771; ZIR2
UniProt ID: (Human) Q96N67
Entrez Gene ID: (Human) 85440
Molecular Function:
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support