Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MDPRGTKRG AEKTEVAEPR NKLPRPAPSL PTDPALYSGP FPFYRRPSEL GCFSLDAQRQ YHGDARALRY YSPPPTNGPG PNFDLRDGYP DRYQPRDEEV QERLDHLLCW LLEHRGRLEG GPGWLAEAIV TWRGHLTKLL TTPYERQEGW QLAASRFQGT LYLSEVETPN ARAQRLARPP LLRELMYMGY KFEQYMCADK PGSSPDPSGE VNTNVAFCSV LRSRLGSHPL LFSGEVDCTD PQAPSTQPPT CYVELKTSKE MHSPGQWRSF YRHKLLKWWA QSFLPGVPNV VAGFRNPDGF VSSLKTFPTM KMFEYVRNDR DGWNPSVCMN FCAAFLSFAQ STVVQDDPRL VHLFSWEPGG PVTVSVHQDA PYAFLPIWYV EAMTQDLPSP PKTPSPK (1-396 aa encoded by BC009344)
This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. The function of its protein product is unknown, but its ubiquitous expression and conservation in both simple and complex eukaryotes suggests that this may be a housekeeping gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-3' exoribonuclease DXO; Decapping and exoribonuclease protein; DeNADding enzyme DXO; Dom-3 homolog Z; DXO; NAD-capped RNA hydrolase DXO; protein Dom3Z
Gene Aliases: DOM3L; DOM3Z; DXO; NG6; RAI1
UniProt ID: (Human) O77932, (Rat) Q6MG77, (Mouse) O70348
Entrez Gene ID: (Human) 1797, (Rat) 361799, (Mouse) 112403
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support