Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of total DPP6 - Catalytic domain.
DPP6 encodes a single-pass type II membrane protein that is a member of the S9B family in clan SC of the serine proteases. This protein has no detectable protease activity, most likely due to the absence of the conserved serine residue normally present in the catalytic domain of serine proteases. However, it does bind specific voltage-gated potassium channels and alters their expression and biophysical properties. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dipeptidyl aminopeptidase IV-related protein; Dipeptidyl aminopeptidase-like protein 6; Dipeptidyl aminopeptidase-related protein; Dipeptidyl peptidase 6; Dipeptidyl peptidase IV-like protein; dipeptidyl peptidase IV-related protein; Dipeptidyl peptidase VI; dipeptidyl-peptidase 6; dipeptidylpeptidase VI; DPP VI; DPPX; In(5)6H p; inversion, Chr 5, Harwell 6, proximal
Gene Aliases: B930011P16Rik; D5Buc3; D5Buc4; D5Buc5; DPL1; DPP VI; Dpp-6; DPP6; DPPX; Gm1377; In(5)6H-p; MRD33; Peplb; Rw; VF2
UniProt ID: (Human) P42658, (Mouse) Q9Z218, (Rat) P46101
Entrez Gene ID: (Human) 1804, (Mouse) 13483, (Rat) 29272
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support