Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RAAYLFIGP SIDVQSFTVH GDSQQLDVFI ATSKHAYATY LWTGEATGQS AFAQVIADRH KILFDRNSAI KSSIVPEVKD YAAIVEQNLQ HFKPVFQLLE KQILSRVRNT ASFRKTAERL LRFSDKRQTE EAIDRIFAIS QQQQQQSKSK KNRRAGKRYK FVDAVPDIFA QIEVNEKKIR QKAQILAQKE LPIDEDEEMK DLLDFADVTY EKHKNGGLIK GRFGQARMVT TTHSRAPSLS ASYTRLFLIL NIAIFFVMLA MQLTYFQRAQ SLHGQRCLYA VLLIDSCILL WLYSSCSQSQ C (659-958 aa encoded by BC039245)
Converts D-glucuronic acid to L-iduronic acid (IdoUA) residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chondroitin-glucuronate 5-epimerase; Dermatan-sulfate epimerase; DS epimerase; SART-2; squamous cell carcinoma antigen recognized by T cells 2; Squamous cell carcinoma antigen recognized by T-cells 2
Gene Aliases: 6030499O08; AI480506; B130024B19Rik; DS-epi1; DSE; DSEP; DSEPI; EDSMC2; SART-2; SART2
UniProt ID: (Human) Q9UL01, (Mouse) Q8BLI4
Entrez Gene ID: (Human) 29940, (Mouse) 212898
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support