Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EDKLILAGNA LQSDSKRLES GVQFQNEAEI AGYILECENL LRQHVIDVQI LIDGKYYQAD QLVQRVAKLR DEIMALRNEC SSVYSKGRIL TTEQTKLMIS
As basal cells of stratified squamous epithelia begin to migrate in response to wound healing, they lose their cell-substrate adhesion junctions, the hemidesmosomes. The hemidesmosome is an adhesion structure of the epidermal-dermal junction in keratinocytes. When keratinocytes migrate laterally or upward to differentiate they must control the formation and disintegration of the hemidesmosomes. The bullous pemphigoid antigen BPAG1 is a hemidesmosomal protein of the cutaneous basement membrane zone. The primary sequence deduced from full-length human cDNAs predicts that this molecule consists of a central rod region and flanking globular domains. A neuronal isoform, BPAG1n3 is the result of differential splicing of BPAG1. BPAG1n3 is distinguished by its initial 32 amino acid residues and by the absence of the amino-terminal half of the actin-binding domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 230 kDa bullous pemphigoid antigen; 230/240 kDa bullous pemphigoid antigen; BPA; Bullous pemphigoid antigen 1; Dystonia musculorum protein; Dystonin; Hemidesmosomal plaque protein; trabeculin-beta
Gene Aliases: BP230; BP240; BPA; BPAG1; CATX-15; CATX15; D6S1101; DMH; DST; DT; EBSB2; HSAN6; KIAA0728; MACF2
UniProt ID: (Human) Q03001
Entrez Gene ID: (Human) 667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support