Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • DTX2 Antibodies

            Abnova

            DTX2 Polyclonal Antibody, MaxPab™

            View all (10) DTX2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite DTX2 Polyclonal Antibody, MaxPab™

            • Antibody Testing Data (1)
            Application
            DTX2 Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            DTX2 Antibody (H00113878-B01P) in WB

            Western Blot analysis of DTX2 expression in transfected 293T cell line by DTX2 MaxPab polyclonal antibody. Lane 1: DTX2 transfected lysate(68.42 KDa). Lane 2: Non-transfected lysate. .
            View Product

            DTX2 Antibody in Western Blot (WB)

            Product Details

            H00113878-B01P

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            DTX2 (NP_065943.1, 1 a.a. approximately 622 a.a) full-length human protein.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.5-1 mg/mL

            Purification

            Protein A

            Storage buffer

            PBS, pH 7.4

            Contains

            no preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            Product may be used with Western Blot (Transfected lysate).

            The Isotype of this product is composed of an IgG Mixture.

            Immunogen sequence: MAMAPSPSLV QVYTSPAAVA VWEWQDGLGT WHPYSATVCS FIEQQFVQQK GQRFGLGSLA HSIPLGQADP SLAPYIIDLP SWTQFRQDTG TMRAVRRHLF PQHSAPGRGV VWEWLSDDGS WTAYEASVCD YLEQQVARGN QLVDLAPLGY NYTVNYTTHT QTNKTSSFCR SVRRQAGPPY PVTTIIAPPG HTGVACSCHQ CLSGSRTGPV SGRYRHSMTN LPAYPVPQHP PHRTASVFGT HQAFAPYNKP SLSGARSAPR LNTTNAWGAA PPSLGSQPLY RSSLSHLGPQ HLPPGSSTSG AVSASLPSGP SSSPGSVPAT VPMQMPKPSR VQQALAGMTS VLMSAIGLPV CLSRAPQPTS PPASRLASKS HGSVKRLRKM SVKGATPKPE PEPEQVIKNY TEELKVPPDE DCIICMEKLS AASGYSDVTD SKAIGSLAVG HLTKCSHAFH LLCLLAMYCN GNKDGSLQCP SCKTIYGEKT GTQPQGKMEV LRFQMSLPGH EDCGTILIVY SIPHGIQGPE HPNPGKPFTA RGFPRQCYLP DNAQGRKVLE LLKVAWKRRL IFTVGTSSTT GETDTVVWNE IHHKTEMDRN ITGHGYPDPN YLQNVLAELA AQGVTEDCLE QQ

            Target Information

            DTX2 functions as an E3 ubiquitin ligase. Regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. Probably acts both as a positive and negative regulator of Notch, depending on the developmental and cell context. Mediates the antineural activity of Notch, possibly by inhibiting the transcriptional activation mediated by MATCH1. Functions as an ubiquitin ligase protein in vitro, suggesting that it may regulate the Notch pathway via some ubiquitin ligase activity.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: deltex 2, E3 ubiquitin ligase; deltex homolog 2; Deltex2; hDTX2; Probable E3 ubiquitin-protein ligase DTX2; Protein deltex-2; RING finger protein 58; RING-type E3 ubiquitin transferase DTX2; zinc ion binding protein

            View more View less

            Gene Aliases: DTX2; KIAA1528; RNF58

            View more View less

            UniProt ID: (Human) Q86UW9

            View more View less

            Entrez Gene ID: (Human) 113878

            View more View less

            Function(s)

            protein binding zinc ion binding ligase activity ubiquitin-protein ligase

            Process(es)

            Notch signaling pathway protein ubiquitination
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-4ftp4:80/100.66.77.180:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0