Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTSGEVKTSL KNAYSSAKRL SPKMEEEGEE EDYCTPGAFE LERLFWKGSP QYTHVNEVWP KLYIGDEATA LDRYRLQKAG FTHVLNAAHG RWNVDTGPDY YRDMDIQYHG VEADDLPTFD LSVFFYPAAA FIDRALSDDH SKILVHCVMG RSRSATLVLA YLMIHKDMTL VDAIQQVAKN RCVLPNRGFL KQLRELDKQL VQQRRRSQRQ DGEEEDGREL
Dual specificity phosphatase DUPD1 is able to dephosphorylate phosphotyrosine, phosphoserine and phosphothreonine residues, with a preference for phosphotyrosine as a substrate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: atypical dual-specific protein phosphatase; Dual specificity phosphatase 27; Dual specificity phosphatase 29; Dual specificity phosphatase DUPD1
Gene Aliases: DUPD1; DUSP27; DUSP29; FMDSP
UniProt ID: (Human) Q68J44
Entrez Gene ID: (Human) 338599
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support