Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MYSLNQEIK AFSRNNLRKQ CTRVTTLTGK KIIETWKDAR IHVVEEVEPS SGGGCGYVQD LSSDLQVGVI KPWLLLGSQD AAHDLDTLKK NKVTHILNVA YGVENAFLSD FTYKSISILD LPETNILSYF PECFEFIEEA KRKDGVVLVH CNAGVSRAAA IVIGFLMNSE QTSFTSAFSL VKNARPSICP NSGFMEQLRT YQEGKESNKC DRIQENSS (1-217 aa encoded by BC035000)
Dual-specificity phosphatases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dual specificity phosphatase TS-DSP1; Dual specificity protein phosphatase 19; LMW-DSP3; Low molecular weight dual specificity phosphatase 3; Protein phosphatase SKRP1; SAPK pathway-regulating phosphatase 1; Stress-activated protein kinase pathway-regulating phosphatase 1
Gene Aliases: 5930436K22Rik; C79103; DUSP17; DUSP19; LMWDSP3; SKRP1; TS-DSP1
UniProt ID: (Human) Q8WTR2
Entrez Gene ID: (Human) 142679, (Mouse) 68082, (Rat) 311151
Molecular Function: protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support