Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAETKIIYHM DEEETPYLVK LPVAPERVTL ADFKNVLSNR PVHAYKFFFK SMDQDFGVVK EEIFDDNAKL PCFNGRVVSW LVLAEGAHSD AGSQGTDSHT DLPPPLERTG GIGDSRPPSF HPNVASSRDG MDNETGTESM VSHRRERARR RNREEAARTN GHPRGDRRRD VGLPPDSAST ALSSELESSS FVDSDEDGST SRLSSSTEQS TSSRLTRKYA SSLLKHGFLR HTVNKITFSE QCYYVFGDLC SNLATLNLNS GSSGTSDQDT LAPLPHPAAP WPLGQGYPYQ YPGPPPCFPP AYQDPGFSYG SGSTGSQQSE GSKSSGSTRS SRRAPGREKE RRAAGAGGSG SESDHTAPSG VGSSWRERPA GQLSRGSSPR SQASATAPGL PPPHPTTKAY TVVGGPPGGP PVRELAAVPP ELTGSRQSFQ KAMGNPCEFF VDIM
DVL1, the human homolog of the Drosophila dishevelled gene (dsh) encodes a cytoplasmic phosphoprotein that regulates cellproliferation, acting as a transducer molecule for developmentalprocesses, including segmentation and neuroblast specification.DVL1 is a candidate gene for neuroblastomatous transformation. TheSchwartz-Jampel syndrome and Charcot-Marie-Tooth disease type 2Ahave been mapped to the same region as DVL1. The phenotypes ofthese diseases may be consistent with defects which might beexpected from aberrant expression of a DVL gene during development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dishevelled 1 (homologous to Drosophila dsh); dishevelled, dsh homolog 1; Dishevelled-1; Dishevelled-1-like; DSH homolog 1; DSH homolog 1-like; MGC54245; RP5-890O3.5; Segment polarity protein dishevelled homolog DVL-1
Gene Aliases: DRS2; DVL; DVL1; DVL1L1; DVL1P1
UniProt ID: (Human) O14640
Entrez Gene ID: (Human) 1855
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support