Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MGETKIIYHL DGQETPYLVK LPLPAERVTL ADFKGVLQRP SYKFFFKSMD DDFGVVKEEI SDDNAKLPCF NGRVVSWLVS AEGSHPDPAP FCADNPSELP
This gene is a member of a multi-gene family which shares strong similarity with the Drosophila disheveled gene, dsh. The Drosophila disheveled gene encodes a cytoplasmic phosphoprotein that regulates cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dishevelled 3 (homologous to Drosophila dsh); dishevelled, dsh homolog 3; Dishevelled-3; DSH homolog 3; dvl-3; OTTHUMP00000210778; Segment polarity protein dishevelled homolog DVL-3
Gene Aliases: DRS3; DVL3; KIAA0208
UniProt ID: (Human) Q92997
Entrez Gene ID: (Human) 1857
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support