Product References
Proteomic Identification of Coxiella burnetii Effector Proteins Targeted to the Host Cell Mitochondria During Infection.
Molecular & cellular proteomics : MCP
Fielden LF,Scott NE,Palmer CS,Khoo CA,Newton HJ,Stojanovski D
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Western Blot
Mon Mar 14 00:00:00 UTC 2022
Proteomic Identification of Coxiella burnetii Effector Proteins Targeted to the Host Cell Mitochondria During Infection.
Molecular & cellular proteomics : MCP
Fielden LF,Scott NE,Palmer CS,Khoo CA,Newton HJ,Stojanovski D
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Western Blot
Mon Mar 14 00:00:00 UTC 2022
Phosphorylation of Connexin36 near the C-terminus switches binding affinities for PDZ-domain and 14-3-3 proteins in vitro.
Scientific reports
Tetenborg S,Wang HY,Nemitz L,Depping A,Espejo AB,Aseervatham J,Bedford MT,Janssen-Bienhold U,O'Brien J,Dedek K
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Immunocytochemistry
Tue Oct 27 00:00:00 UTC 2020
An efficient and quantitative assay for epitope-tagged therapeutic protein development with a capillary western system.
Bioanalysis
Li Y,Duah E,Long N,Persaud A,VanGundy Z,Magliery T,Poi MJ
PA1-984B-HRP was used in Simple Western - Size to develop and validate a reliable, robust and efficient assay to detect and quantify biologic compounds in vitro and in vivo during early stage of a biotherapeutic agent discovery.
Fri Mar 01 00:00:00 UTC 2019
Identification and Characterization of USP7 Targets in Cancer Cells.
Scientific reports
Georges A,Marcon E,Greenblatt J,Frappier L
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Western Blot
Fri Oct 26 00:00:00 UTC 2018
Novel antibodies reveal presynaptic localization of C9orf72 protein and reduced protein levels in C9orf72 mutation carriers.
Acta neuropathologica communications
Frick P,Sellier C,Mackenzie IRA,Cheng CY,Tahraoui-Bories J,Martinat C,Pasterkamp RJ,Prudlo J,Edbauer D,Oulad-Abdelghani M,Feederle R,Charlet-Berguerand N,Neumann M
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Western Blot
Fri Aug 03 00:00:00 UTC 2018
Methylation of the phosphatase-transcription activator EYA1 by protein arginine methyltransferase 1: mechanistic, functional, and structural studies.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Li X,Eberhardt A,Hansen JN,Bohmann D,Li H,Schor NF
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Immunoprecipitation
Thu Jun 01 00:00:00 UTC 2017
Selective regulation of Mmp13 by 1,25(OH)2D3, PTH, and Osterix through distal enhancers.
The Journal of steroid biochemistry and molecular biology
Meyer MB,Benkusky NA,Onal M,Pike JW
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in ChIP assay
Tue Nov 01 00:00:00 UTC 2016
Selective regulation of Mmp13 by 1,25(OH)2D3, PTH, and Osterix through distal enhancers.
The Journal of steroid biochemistry and molecular biology
Meyer MB,Benkusky NA,Onal M,Pike JW
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in ChIP assay
Tue Nov 01 00:00:00 UTC 2016
The production of viral vectors designed to express large and difficult to express transgenes within neurons.
Molecular brain
Holehonnur R,Lella SK,Ho A,Luong JA,Ploski JE
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Immunofluorescence
Tue Feb 24 00:00:00 UTC 2015
The production of viral vectors designed to express large and difficult to express transgenes within neurons.
Molecular brain
Holehonnur R,Lella SK,Ho A,Luong JA,Ploski JE
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Immunofluorescence
Tue Feb 24 00:00:00 UTC 2015
Distinct roles of neuroligin-1 and SynCAM1 in synapse formation and function in primary hippocampal neuronal cultures.
Neuroscience
Burton SD,Johnson JW,Zeringue HC,Meriney SD
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Western Blot
Thu Jul 26 00:00:00 UTC 2012
Virus-infection or 5'ppp-RNA activates antiviral signal through redistribution of IPS-1 mediated by MFN1.
PLoS pathogens
Onoguchi K,Onomoto K,Takamatsu S,Jogi M,Takemura A,Morimoto S,Julkunen I,Namiki H,Yoneyama M,Fujita T
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Western Blot
Thu Jul 22 00:00:00 UTC 2010
Virus-infection or 5'ppp-RNA activates antiviral signal through redistribution of IPS-1 mediated by MFN1.
PLoS pathogens
Onoguchi K,Onomoto K,Takamatsu S,Jogi M,Takemura A,Morimoto S,Julkunen I,Namiki H,Yoneyama M,Fujita T
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Western Blot
Thu Jul 22 00:00:00 UTC 2010
Virus-infection or 5'ppp-RNA activates antiviral signal through redistribution of IPS-1 mediated by MFN1.
PLoS pathogens
Onoguchi K,Onomoto K,Takamatsu S,Jogi M,Takemura A,Morimoto S,Julkunen I,Namiki H,Yoneyama M,Fujita T
Published figure using DYKDDDDK Tag polyclonal antibody (Product # PA1-984B-HRP) in Western Blot
Thu Jul 22 00:00:00 UTC 2010