Product References
NaV1.6 dysregulation within myocardial T-tubules by D96V calmodulin enhances proarrhythmic sodium and calcium mishandling.
The Journal of clinical investigation
Tarasov M,Struckman HL,Olgar Y,Miller A,Demirtas M,Bogdanov V,Terentyeva R,Soltisz AM,Meng X,Min D,Sakuta G,Dunlap I,Duran AD,Foster MP,Davis JP,Terentyev D,Györke S,Veeraraghavan R,Radwański PB
MA1-142-A488 was used in Immunohistochemistry (PFA fixed) to demonstrate that D96V-CaM promotes arrhythmias by dysregulating LTCC and NaV1.6 within T-tubules and thereby, facilitating aberrant Ca2+ release.
Mon Apr 03 00:00:00 UTC 2023
GAPDH mediates plant reovirus-induced incomplete autophagy for persistent viral infection in leafhopper vector.
Autophagy
Chen Q,Zhang Y,Yang H,Wang X,Ren J,Jia D,Chen H,Wei T
MA1-142-A488 was used in Immunocytochemistry to reveal a positive regulation of immune response in insect vectors during viral infection.
Sat Apr 01 00:00:00 UTC 2023
An integrated model for Gpr124 function in Wnt7a/b signaling among vertebrates.
Cell reports
America M,Bostaille N,Eubelen M,Martin M,Stainier DYR,Vanhollebeke B
MA1-142-A488 was used in Immunocytochemistry, Western Blot, Immunoprecipitation to provide insights into the evolution of Gpr124/Reck function across the vertebrate clade, a receptor complex uniquely implicated in Wnt ligand-specific cellular responses.
Tue May 31 00:00:00 UTC 2022
Rapid generation of potent antibodies by autonomous hypermutation in yeast.
Nature chemical biology
Wellner A,McMahon C,Gilman MSA,Clements JR,Clark S,Nguyen KM,Ho MH,Hu VJ,Shin JE,Feldman J,Hauser BM,Caradonna TM,Wingler LM,Schmidt AG,Marks DS,Abraham J,Kruse AC,Liu CC
MA1-142-A488 was used in Flow Cytometry to develop the autonomous hypermutation yeast surface display technique for recombinant antibody generation.
Fri Oct 01 00:00:00 UTC 2021
High-purity production and precise editing of DNA base editing ribonucleoproteins.
Science advances
Jang HK,Jo DH,Lee SN,Cho CS,Jeong YK,Jung Y,Yu J,Kim JH,Woo JS,Bae S
MA1-142-A488 was used in Immunohistochemistry to applied NG PAM-targetable ABE RNPs to in vivo gene correction in retinal degeneration 12 (rd12) model mice.
Sun Aug 01 00:00:00 UTC 2021
M448R and MGF505-7R: Two African Swine Fever Virus Antigens Commonly Recognized by ASFV-Specific T-Cells and with Protective Potential.
Vaccines
Bosch-Camós L,López E,Collado J,Navas MJ,Blanco-Fuertes M,Pina-Pedrero S,Accensi F,Salas ML,Mundt E,Nikolin V,Rodríguez F
MA1-142-A488 was used in Immunocytochemistry-immunofluorescence to use data from the SLAI-peptide repertoire presented by a single set of african swine fever virus-infected porcine alveolar macrophages to generate a complex DNA vaccine composed by 15 plasmids encoding the individual peptide-bearing ORFs.
Fri May 14 00:00:00 UTC 2021
Oxidative pentose phosphate pathway and glucose anaplerosis support maintenance of mitochondrial NADPH pool under mitochondrial oxidative stress.
Bioengineering & translational medicine
Moon SJ,Dong W,Stephanopoulos GN,Sikes HD
Published figure using DYKDDDDK Tag monoclonal antibody (Product # MA1-142-A488) in Immunocytochemistry
Tue Sep 01 00:00:00 UTC 2020
Short Stature is Progressive in Patients with Heterozygous NPR2 Mutations.
The Journal of clinical endocrinology and metabolism
Hanley PC,Kanwar HS,Martineau C,Levine MA
MA1-142-A488 was used in Immunocytochemistry to show that iallelic NPR2 mutations cause severe skeletal dysplasia (AMDM), whereas heterozygous mutations lead to a subtler phenotype characterized by progressive short stature with by increasing loss of height potential with age.
Thu Oct 01 00:00:00 UTC 2020
Structural and functional characterization of G protein-coupled receptors with deep mutational scanning.
eLife
Jones EM,Lubock NB,Venkatakrishnan AJ,Wang J,Tseng AM,Paggi JM,Latorraca NR,Cancilla D,Satyadi M,Davis JE,Babu MM,Dror RO,Kosuri S
MA1-142-A488 was used in Immunocytochemistry to characterize large libraries of GPCR variants in human cell lines with a barcoded transcriptional reporter of G protein signal transduction.
Wed Oct 21 00:00:00 UTC 2020
Raptor-Mediated Proteasomal Degradation of Deamidated 4E-BP2 Regulates Postnatal Neuronal Translation and NF-κB Activity.
Cell reports
Kouloulia S,Hallin EI,Simbriger K,Amorim IS,Lach G,Amvrosiadis T,Chalkiadaki K,Kampaite A,Truong VT,Hooshmandi M,Jafarnejad SM,Skehel P,Kursula P,Khoutorsky A,Gkogkas CG
Published figure using DYKDDDDK Tag monoclonal antibody (Product # MA1-142-A488) in Immunocytochemistry
Tue Dec 10 00:00:00 UTC 2019
In vivo stabilization of OPA1 in hepatocytes potentiates mitochondrial respiration and gluconeogenesis in a prohibitin-dependent way.
The Journal of biological chemistry
Li L,Martin-Levilain J,Jiménez-Sánchez C,Karaca M,Foti M,Martinou JC,Maechler P
Published figure using DYKDDDDK Tag monoclonal antibody (Product # MA1-142-A488) in Immunofluorescence
Fri Aug 23 00:00:00 UTC 2019
In vivo stabilization of OPA1 in hepatocytes potentiates mitochondrial respiration and gluconeogenesis in a prohibitin-dependent way.
The Journal of biological chemistry
Li L,Martin-Levilain J,Jiménez-Sánchez C,Karaca M,Foti M,Martinou JC,Maechler P
Published figure using DYKDDDDK Tag monoclonal antibody (Product # MA1-142-A488) in Immunofluorescence
Fri Aug 23 00:00:00 UTC 2019
In vivo stabilization of OPA1 in hepatocytes potentiates mitochondrial respiration and gluconeogenesis in a prohibitin-dependent way.
The Journal of biological chemistry
Li L,Martin-Levilain J,Jiménez-Sánchez C,Karaca M,Foti M,Martinou JC,Maechler P
Published figure using DYKDDDDK Tag monoclonal antibody (Product # MA1-142-A488) in Immunofluorescence
Fri Aug 23 00:00:00 UTC 2019