Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • DYNC1H1 Antibodies

            Invitrogen

            DYNC1H1 Polyclonal Antibody

            Advanced Verification
            2 References
            View all (9) DYNC1H1 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # PA5-49451
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (JPY)
            53,900
            Online offer
            Ends: 26-Sep-2025
            84,000
            Save 30,100 (36%)
            400 µL
             
            Contact Us
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite DYNC1H1 Polyclonal Antibody

            • Antibody Testing Data (4)
            • Advanced Verification (1)
            Application
            DYNC1H1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 5

            DYNC1H1 Antibody (PA5-49451) in IHC (P)

            Immunohistochemistry analysis of DYNC1H1 in paraformaldehyde-fixed, paraffin-embedded human testis sections. Samples were incubated with DYNC1H1 polyclonal antibody (Product # PA5-49451) using a dilution of 1:25 for 1 hours at 37°C followed by an undiluted biotinylated goat polyvalent antibody. Tissue was fixed with formaldehyde and blocked with 3% BSA for 0.5 hour at room temperature; antigen retrieval was... View More
            View Product

            DYNC1H1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            DYNC1H1 Antibody in Western Blot (WB)
            DYNC1H1 Antibody in Western Blot (WB)
            DYNC1H1 Antibody in Western Blot (WB)
            DYNC1H1 Antibody

            Product Details

            PA5-49451

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:2,000
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:25
            -

            In Situ Hybridization (ISH)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human, Mouse

            Published species

            Human

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            KLH conjugated synthetic peptide between 4202-4236 amino acids from the C-terminal region of human DYNC1H1.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.5 mg/mL

            Purification

            Protein A, Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.4

            Contains

            0.09% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2634905

            Target Information

            DYNC1H1 (Dynein Cytoplasmic 1 Heavy Chain 1) is a gene that encodes a protein belonging to the dynein family, which are motor proteins critical for various cellular processes. Dyneins are responsible for retrograde intracellular transport along microtubules, meaning they move cargo from the periphery of the cell towards the nucleus. The DYNC1H1 protein is a heavy chain component of cytoplasmic dynein 1, which plays a key role in the movement of organelles, vesicles, and other cellular components. Mutations in DYNC1H1 have been associated with several neurological disorders, including spinal muscular atrophy with lower extremity predominance (SMA-LED) and Charcot-Marie-Tooth disease type 2O (CMT2O). Research into DYNC1H1 continues to uncover its involvement in cellular mechanics and its implications in various genetic conditions.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: Cytoplasmic dynein 1 heavy chain 1; Cytoplasmic dynein heavy chain 1; Dynein heavy chain, cytosolic; dynein heavy chain, retrograde transport; dynein, cytoplasmic 1, heavy chain 1; dynein, cytoplasmic, heavy chain 1; dynein, cytoplasmic, heavy polypeptide 1; legs at odd angle; sprawling

            View more View less

            Gene Aliases: 9930018I23Rik; AI894280; CMT2O; DHC1; DHC1a; DNCH1; Dnchc1; DNCL; Dnec1; DNECL; DYHC; DYNC1H1; HL-3; KIAA0325; Loa; MAP1C; mKIAA0325; P22; SMALED1; Swl

            View more View less

            UniProt ID: (Human) Q14204, (Mouse) Q9JHU4

            View more View less

            Entrez Gene ID: (Human) 1778, (Mouse) 13424

            View more View less

            Function(s)

            microtubule motor activity protein binding ATP binding ATPase activity poly(A) RNA binding dynein light intermediate chain binding nucleotide binding motor activity microtubule binding motor protein cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein

            Process(es)

            G2/M transition of mitotic cell cycle ER to Golgi vesicle-mediated transport microtubule-based movement mitotic spindle organization antigen processing and presentation of exogenous peptide antigen via MHC class II cytoplasmic mRNA processing body assembly stress granule assembly establishment of spindle localization cilium movement transport
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            japan | 日本 flag icon
            Japan | 日本

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0