Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: FLDDGNQMLL RVQRSDAGIS FSNTIEFGDT KDKVLVFFKL RPEVITDENL HDNILVSSML ESPISSLYQA VRQVFAPMLL KDQEWSRNFD PKLQNLLSEL EAGLGIVLRR SDTNLTKLKF KEDDTRGILT PSDEFQFWIE QAHRGNKQIS KERAN
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic dynein 2 heavy chain; Cytoplasmic dynein 2 heavy chain 1; cytoplasmic dynein heavy chain 2; Dynein cytoplasmic heavy chain 2; Dynein heavy chain 11; dynein heavy chain 2; Dynein heavy chain isotype 1B; dynein heavy chain, isotype 1B; dynein, cytoplasmic 2, heavy chain 1; dynein, cytoplasmic, heavy chain 2; dynein, cytoplasmic, heavy polypeptide 2; hDHC11; mDHC11
Gene Aliases: 4432416O06Rik; AI448217; ATD3; b2b414Clo; D030010H02Rik; D330044F14Rik; DHC1B; DHC2; DNCH2; Dnchc2; DYH1B; DYNC2H1; hdhc11; KIAA1997; mKIAA1997; SRPS2B; SRTD3
UniProt ID: (Human) Q8NCM8, (Mouse) Q45VK7
Entrez Gene ID: (Human) 79659, (Mouse) 110350
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support