Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: A549, H1299, MCF-7.
Predicted reactivity: Human (99%), Mouse (87%), Rat (88%), Xenopus laevis (80%), Chicken (82%), Chimpanzee (99%), Bovine (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a tetratricopeptide repeat domain-containing protein. The encoded protein interacts with estrogen receptors and the heat shock proteins, Hsp70 and Hsp90. A chromosomal translocation involving this gene is associated with a susceptibility to developmental dyslexia. Mutations in this gene are associated with deficits in reading and spelling. Alternate splicing results in multiple transcript variants.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dynein axonemal assembly factor 4; dynein, axonemal, assembly factor 4; Dyslexia susceptibility 1 candidate gene 1 protein; FLJ37882; MGC70618
Gene Aliases: CILD25; DNAAF4; DYX1; DYX1C1; DYXC1; EKN1; RD
UniProt ID: (Human) Q8WXU2
Entrez Gene ID: (Human) 161582
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support