Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Specificity: Human DZIP1
May participate in spermatogenesis via its interaction with DAZ.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilium assembly protein DZIP1; DAZ interacting protein 1; DAZ interacting protein testis1; DAZ-interacting protein 1/2; DAZ-interacting zinc finger protein 1; zinc finger DAZ interacting protein 1; zinc-finger protein DZIPt1
Gene Aliases: DZIP; DZIP1; DZIP2; DZIPt1; KIAA0996
UniProt ID: (Human) Q86YF9
Entrez Gene ID: (Human) 22873
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support