Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: Dectin-2.
Dectin-2 is a C-type lectin expressed by dendritic cells, macrophages and inflammatory monocytes. It is a type II membrane receptor, with a short cytoplasmic tail, and a C-terminal C-type lectin domain. Dectin-2 is a receptor with specificity for high mannose structures and mediates cytokine production from DC and other antigen presenting cells. Though, Dectin-2 has no obvious signaling motifs attached to CP tail, reports indicate that Dectin-2 employs FcRg for internalization and to activate NF-kappaB pathway for cytokine production. Though it shares only 20-25% of homology with Dectin-1, it functions similarly as pattern recognition receptor (PRR) for fungal infections, to activate both innate (Syk-CARD9 innate signaling pathway) and adaptive immune responses (by activation of DC) to fungal infections. Thus Dectin-2 is a connector molecule of both innate and adaptive immunity functioning at DC level.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 10; C-type lectin domain family 6 member A; C-type lectin domain family 6, member A; C-type lectin superfamily member 10; DC-associated C-type lectin 2; dectin 2; dectin-2; Dendritic cell-associated C-type lectin 2
Gene Aliases: CLEC4N; CLEC6A; CLECSF10; DECTIN2
UniProt ID: (Human) Q6EIG7
Entrez Gene ID: (Human) 93978
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support