Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: EPLQARADEV AAAPEQIAAD IPEVVVSLAW DESLAPKHPG SRKNMDCYCR IPACIAGERR YGTCIYQGRL WAFCC; Positive Samples: MCF-7, 22Rv1, SW480
Defensin alpha-1 is part of a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. Defensins are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The Defensin alpha-1 protein is encoded by the DFA1 gene and is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8, and this gene differs from defensin, alpha 3 by only one amino acid. The DFA1 gene and the gene encoding defensin, alpha 3 are both subject to copy number variation. Two transcript variants encoding different isoforms of Defensin alpha-1 have been found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: defensin 3, neutrophil-specific; Defensin, alpha 3; defensin, alpha 3, neutrophil-specific; HNP-3; HP-3; Neutrophil defensin 3; neutrophil peptide 3
Gene Aliases: DEF3; DEFA3; HNP-3; HNP3; HP-3; HP3
UniProt ID: (Human) P59666
Entrez Gene ID: (Human) 1668
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support