Search Thermo Fisher Scientific
FIGURE: 1 / 1
Studies in Drosophila have identified this gene as encoding a positive regulator of the Notch-signaling pathway. The human gene encodes a protein of unknown function; however, it may play a role in basic helix-loop-helix transcription factor activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: deltex 1; deltex 1, E3 ubiquitin ligase; deltex homolog 1; Deltex1; E3 ubiquitin-protein ligase DTX1; fractionated x-irradiation induced transcript 1; FXI-T1; hDTX1; mDTX1; Protein deltex-1; RING-type E3 ubiquitin transferase DTX1
Gene Aliases: DTX1; Fxit1; hDx-1; mKIAA4160
UniProt ID: (Human) Q86Y01, (Mouse) Q61010
Entrez Gene ID: (Human) 1840, (Rat) 687424, (Mouse) 14357
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support