Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute lyophilized product with 1 mL distilled water.
DG 3.10 recognizes desmoglein 1 + 2 (desmosome-specific cadherins) in desmosome-bearing cells; does not react with cells containing plaque-bearing junctions of non-desmosomal type. Tumors specifically detected: all epithelium-derived tumors, notably carcinomas, and meningiomas. Polypeptide reacting: MW 165 000 Desmoglein, "band 3" polypeptide of the desmosomal complex. Epitope recognized has been localized at the repeating unit domain ("RUD") of the cytoplasmic C-terminal part of the polypeptide.Reactivity on cultured cell lines: A-431, MDBK.
DSG1 (Desmoglein 1) is a Protein Coding gene. Diseases associated with DSG1 include Erythroderma, Congenital, With Palmoplantar Keratoderma, Hypotrichosis, And Hyper-Ige and Palmoplantar Keratoderma I, Striate, Focal, Or Diffuse. Among its related pathways are Keratinization and Apoptotic execution phase. Gene Ontology (GO) annotations related to this gene include calcium ion binding and toxic substance binding. An important paralog of this gene is DSG4. DSG2 (Desmoglein 2) is a Protein Coding gene. Diseases associated with DSG2 include Arrhythmogenic Right Ventricular Dysplasia, Familial, 10 and Cardiomyopathy, Dilated, 1Bb. Among its related pathways are Keratinization and Apoptotic execution phase. Gene Ontology (GO) annotations related to this gene include calcium ion binding and cell adhesive protein binding involved in bundle of His cell-Purkinje myocyte communication. An important paralog of this gene is DSG4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cadherin family member 4; Cadherin family member 5; desmoglein 1; desmoglein 1 beta; desmoglein 1 gamma; desmoglein 2; Desmoglein-1; Desmoglein-2; Desmosomal glycoprotein 1; DG1; DGI; DSG1; DSG1/2; DSG2; HDGC; Pemphigus foliaceus antigen
Gene Aliases: CDHF4; CDHF5; DG1; DSG; DSG1; Dsg1b; Dsg1c; DSG2; EPKHE; EPKHIA; HDGC; PPKS1; SPPK1
UniProt ID: (Human) Q02413, (Bovine) Q03763, (Human) Q14126
Entrez Gene ID: (Human) 1828, (Bovine) 281131, (Human) 1829, (Bovine) 508151, (Rat) 291755, (Rat) 307562
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support