Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Specificity: desmoglein 2 present in all epithelia and in tumors derived therefrom as well as in heart muscle and dendritic reticulum cells of lymphatic follicles. In basal epithelial cells desmoglein 2 expression is generally very low. Polypeptide reacting: Desmoglein 2 (1069aa; calculated mol. wt. 116,760).Recommended as positive control: CaCo-2, HaCaT.Reactivity on cultured cell lines: Many, including CaCo-2, HaCaT, MCF-7. This antibody was used in a Indirect ELISA.
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARVC10; ARVD10; Cadherin family member 5; CMD1BB; desmoglein 2; Desmoglein-2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037
Gene Aliases: CDHF5; DSG2; HDGC
UniProt ID: (Human) Q14126
Entrez Gene ID: (Human) 1829, (Rat) 307562
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support