Product References
Differentiation of Human Induced Pluripotent Stem Cells into Keratinocytes.
Current protocols
Koch PJ,Webb S,Gugger JA,Salois MN,Koster MI
MA5-16025 was used in Dot Blot, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to provide the basic protocol for the generation of human iPSC-derived keratinocytes (iPSC-K).
Fri Apr 01 00:00:00 EDT 2022
Differentiation of Human Induced Pluripotent Stem Cells into Keratinocytes.
Current protocols
Koch PJ,Webb S,Gugger JA,Salois MN,Koster MI
MA5-16025 was used in Dot Blot, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to provide the basic protocol for the generation of human iPSC-derived keratinocytes (iPSC-K).
Fri Apr 01 00:00:00 EDT 2022
A non-canonical role for desmoglein-2 in endothelial cells: implications for neoangiogenesis.
Angiogenesis
Ebert LM,Tan LY,Johan MZ,Min KK,Cockshell MP,Parham KA,Betterman KL,Szeto P,Boyle S,Silva L,Peng A,Zhang Y,Ruszkiewicz A,Zannettino AC,Gronthos S,Koblar S,Harvey NL,Lopez AF,Shackleton M,Bonder CS
MA516025 was used in flow cytometry to study the function of desmoglein-2 and generate knockout mice
Sat Oct 01 00:00:00 EDT 2016
De novo epidermal regeneration using human eccrine sweat gland cells: higher competence of secretory over absorptive cells.
The Journal of investigative dermatology
Pontiggia L,Biedermann T,Böttcher-Haberzeth S,Oliveira C,Braziulis E,Klar AS,Meuli-Simmen C,Meuli M,Reichmann E
MA5-16025 was used in immunohistochemistry to study the role of human eccrine sweat gland cells during epidermal regeneration
Sun Jun 01 00:00:00 EDT 2014
MAPKAP kinase 2 (MK2)-dependent and -independent models of blister formation in pemphigus vulgaris.
The Journal of investigative dermatology
Mao X,Li H,Sano Y,Gaestel M,Mo Park J,Payne AS
MA5-16025 was used in immunocytochemistry and western blot to identify MK2-dependent and -independent mechanisms of blister formation in a mouse model of pemphigus vulgaris
Wed Jan 01 00:00:00 EST 2014
Altered expression and localization of desmoglein 3 in esophageal squamous cell carcinoma.
Acta histochemica
Fang WK,Chen B,Xu XE,Liao LD,Wu ZY,Wu JY,Shen J,Xu LY,Li EM
MA5-16025 was used in immunohistochemistry - paraffin section to examine desmoglein 3 expression and subcellular localization in esophageal squamous cell carcinoma.
Sun Jun 01 00:00:00 EDT 2014
Desmoglein 2 compensates for desmoglein 3 but does not control cell adhesion via regulation of p38 mitogen-activated protein kinase in keratinocytes.
The Journal of biological chemistry
Hartlieb E,Rötzer V,Radeva M,Spindler V,Waschke J
Published figure using Desmoglein 3 monoclonal antibody (Product # MA5-16025) in Immunoprecipitation
Fri Jun 13 00:00:00 EDT 2014
Desmoglein 2 compensates for desmoglein 3 but does not control cell adhesion via regulation of p38 mitogen-activated protein kinase in keratinocytes.
The Journal of biological chemistry
Hartlieb E,Rötzer V,Radeva M,Spindler V,Waschke J
Published figure using Desmoglein 3 monoclonal antibody (Product # MA5-16025) in Immunoprecipitation
Fri Jun 13 00:00:00 EDT 2014
Desmoglein 2 compensates for desmoglein 3 but does not control cell adhesion via regulation of p38 mitogen-activated protein kinase in keratinocytes.
The Journal of biological chemistry
Hartlieb E,Rötzer V,Radeva M,Spindler V,Waschke J
Published figure using Desmoglein 3 monoclonal antibody (Product # MA5-16025) in Immunoprecipitation
Fri Jun 13 00:00:00 EDT 2014
Adducin is required for desmosomal cohesion in keratinocytes.
The Journal of biological chemistry
Rötzer V,Breit A,Waschke J,Spindler V
MA5-16025 was used in immunocytochemistry to study the role of actin-binding protein adducin in desmosomal cohesion in keratinocytes
Fri May 23 00:00:00 EDT 2014
Desmosome assembly and disassembly are membrane raft-dependent.
PloS one
Stahley SN,Saito M,Faundez V,Koval M,Mattheyses AL,Kowalczyk AP
MA5-16025 was used in western blot to investigate the role of membrane rafts during desmosome assembly and disassembly and cell adhesion
Mon Sep 08 00:00:00 EDT 2014
p38 MAPK activation is downstream of the loss of intercellular adhesion in pemphigus vulgaris.
The Journal of biological chemistry
Mao X,Sano Y,Park JM,Payne AS
MA5-16025 was used in immunocytochemistry and immunohistochemistry to investigate the mechanisms by which autoantibodies contribute to pemphigus vulgaris
Fri Jan 14 00:00:00 EST 2011
p38 MAPK activation is downstream of the loss of intercellular adhesion in pemphigus vulgaris.
The Journal of biological chemistry
Mao X,Sano Y,Park JM,Payne AS
MA5-16025 was used in immunocytochemistry and immunohistochemistry to investigate the mechanisms by which autoantibodies contribute to pemphigus vulgaris
Fri Jan 14 00:00:00 EST 2011
Autoimmunity to desmocollin 3 in pemphigus vulgaris.
The American journal of pathology
Mao X,Nagler AR,Farber SA,Choi EJ,Jackson LH,Leiferman KM,Ishii N,Hashimoto T,Amagai M,Zone JJ,Payne AS
MA5-16025 was used in immunocytochemistry, immunohistochemistry, and western blot to report that desmocollin 3 is an autoantigen in pemphigus vulgaris
Wed Dec 01 00:00:00 EST 2010
Desmoglein 2-mediated adhesion is required for intestinal epithelial barrier integrity.
American journal of physiology. Gastrointestinal and liver physiology
Schlegel N,Meir M,Heupel WM,Holthöfer B,Leube RE,Waschke J
MA5-16025 was used in immunocytochemistry to investigate the role of desmosomal adhesion in the intestinal epithelial barrier function
Sat May 01 00:00:00 EDT 2010
Disruption of desmosome assembly by monovalent human pemphigus vulgaris monoclonal antibodies.
The Journal of investigative dermatology
Mao X,Choi EJ,Payne AS
Published figure using Desmoglein 3 monoclonal antibody (Product # MA5-16025) in Immunocytochemistry
Wed Apr 01 00:00:00 EDT 2009
Disruption of desmosome assembly by monovalent human pemphigus vulgaris monoclonal antibodies.
The Journal of investigative dermatology
Mao X,Choi EJ,Payne AS
Published figure using Desmoglein 3 monoclonal antibody (Product # MA5-16025) in Immunocytochemistry
Wed Apr 01 00:00:00 EDT 2009
P120 catenin is associated with desmogleins when desmosomes are assembled in high-Ca2+ medium but not when disassembled in low-Ca2+ medium in DJM-1 cells.
The Journal of dermatology
Kanno M,Aoyama Y,Isa Y,Yamamoto Y,Kitajima Y
MA5-16025 was used in immunoprecipitation to test if p120ctn associates with Dsg1 and Dsg3 in keratinocytes
Sun Jun 01 00:00:00 EDT 2008
Characterization of extracellular matrix components in the limbal epithelial stem cell compartment.
Experimental eye research
Schlötzer-Schrehardt U,Dietrich T,Saito K,Sorokin L,Sasaki T,Paulsson M,Kruse FE
MA5-16025 was used in immunohistochemistry - frozen section to investigate basement membrane components in the limbal epithelial stem cell compartment by immunohistochemistry
Sat Dec 01 00:00:00 EST 2007
New skin-equivalent model from de-epithelialized amnion membrane.
Cell and tissue research
Yang L,Shirakata Y,Shudou M,Dai X,Tokumaru S,Hirakawa S,Sayama K,Hamuro J,Hashimoto K
MA5-16025 was used in immunohistochemistry - paraffin section to develop a living skin-equivalent from de-epithelialized amnion membrane
Sun Oct 01 00:00:00 EDT 2006
Expression of desmosomal proteins in squamous cell carcinomas of the skin.
Journal of cutaneous pathology
Kurzen H,Münzing I,Hartschuh W
MA5-16025 was used in immunohistochemistry - paraffin section to discuss the role of desmosomal proteins to cancer
Sat Nov 01 00:00:00 EST 2003