Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: YPMVEYCTPT TSGEDVRDFA KVLKNKFRTK RYFAKHPRMG YLPVQTVLEG DNMETPVTLI NFWPVDSAPA SSPQLSHDDT HSRIEHYASR LAEMENSNGS YLNDSISPNE SIDDEHLLIQ HYCQSLNQDS PLSQPRSPAQ ILISLESEER GELERILADL EEENRNLQAE YDRLKQQHEH KGLSPLPSPP EMMPTSPQSP RDAELIAEAK LLRQHKGRLE ARMQILEDHN KQLESQLHRL RQLLEQPQAE AKVNGTTVSS PSTSLQRSDS SQPMLLRVVG SQTSDSMGEE DLLSPPQDTS TGLEEVMEQL NNSFPSSRGH NVGSLFHMAD DLGRAMESLV SVMTDEEGAE
Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immunohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: apodystrophin-3; apodystrophin-I; Duchenne muscular dystrophy (DMD); Dystrophin; dystrophin Dp40; dystrophin Dp71a; dystrophin Dp71ab; dystrophin Dp71b; dystrophin, muscular dystrophy; GS1-19O24.1; Muscular dystrophy Duchenne and Becker types; OTTHUMP00000215592; X-linked muscular dystrophy
Gene Aliases: BMD; CMD3B; DMD; DNADMD1; Dp427; Dp71; DXS142; DXS164; DXS206; DXS230; DXS239; DXS268; DXS269; DXS270; DXS272; DXSmh7; DXSmh9; dys; mdx; MRX85; pke
UniProt ID: (Human) P11532, (Mouse) P11531
Entrez Gene ID: (Human) 1756, (Mouse) 13405, (Rat) 24907
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support