Find the ideal FBS using our iMATCH sera lot matching tool

Learn more

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • E-cadherin Antibodies

            Invitrogen

            E-cadherin Monoclonal Antibody (ECCD-1)

            1 Published Figure
            15 References
            View all (173) E-cadherin antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # 13-1800
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            590.00
            100 µg
             
            In stock
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite E-cadherin Monoclonal Antibody (ECCD-1)

            • Published Figures (1)
            Application
            E-cadherin Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            E-cadherin Antibody (13-1800) in WB

            Frontiers in molecular neuroscience 2022 - No change was found in the protein expression level of G-actin. Protein expression level of Cx26, beta-actin (A) , and gamma-actin (E) in P0 control and P0 KD groups. beta-actin and gamma-actin protein expression level in cochlear of Cx26 KD mouse models. The ratio of G-actin/F-actin in P0 control and P0 KD group (B) respectively. Western blotting F-actin and G-actin in Western blot results of protein expre... View More
            Published figure supplied by benchsci-logo
            PMID: 35345836
            View Product

            E-cadherin Antibody in Western Blot (WB)

            Product Details

            13-1800

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            -
            View 4 publications 4 publications

            Immunohistochemistry (IHC)

            -
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            -
            View 1 publication 1 publication

            Immunohistochemistry (Frozen) (IHC (F))

            -
            View 1 publication 1 publication

            Immunocytochemistry (ICC/IF)

            -
            View 3 publications 3 publications

            Neutralization (Neu)

            -
            View 3 publications 3 publications

            Inhibition Assays (IA)

            200 µg/mL
            -

            Miscellaneous PubMed (Misc)

            -
            View 2 publications 2 publications
            Product Specifications

            Species Reactivity

            Mouse

            Published species

            Fish, Human, Mouse, Rat, Tag

            Host/Isotype

            Rat / IgG2b

            Class

            Monoclonal

            Type

            Antibody

            Clone

            ECCD-1

            Immunogen

            Mouse teratocarcinoma cell line F9.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Concentration

            2.0 mg/mL

            Purification

            purified

            Storage buffer

            PBS, pH 7.4, with 1% BSA

            Contains

            no preservative

            Storage conditions

            4°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2533004

            Product Specific Information

            Source: Monoclonal antibody was obtained by fusing the P3-X63-Ag8 mouse myeloma cell line with spleen cells of Wistar rat after immunization with mouse teratocarcinoma cell line F9.

            Specificity: This antibody specifically reacts with mouse E-cadherin. This antibody inhibits E-cadherin dependent cell-cell contact.

            Cross reactivity: This antibody does not react with human, bovine, chicken or dog E-cadherin.

            Storage: 4°C This product does not contain preservative. The stock solution (2.0 mg/mL) should be stored in aliquots at -20°C for 1 year, or should be stored at 4°C for 6 months after adding 0.1% sodium azide. Avoid repeated freeze-thaw cycles.

            Reconstitution: Dissolve the lyophilized antibody in 50 µL of distilled water (final concentration: 2.0 mg/mL). This solution can be used as a stock solution. If dilution is necessary for your application, dilute the stock solution with the following Dilution solution just prior to use. When the entire amount of antibody is to be used over a short time period, it may be dissolved directly in 500 µL or more of the Dilution solution. Be sure to store the antibody at a minimum concentration of 2.0 mg/mL. A lower antibody concentration may result in decreased stability. Reconstituted antibody solution should contain 0.1% sodium azide as a preservative when stored at 4°C.

            Target Information

            E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintenance of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: ARC-1; cadherin e; Cadherin-1; cadherin-E; CD324; E-cadherin; Ecadherin; Epithelial cadherin; Uvomorulin

            View more View less

            Gene Aliases: AA960649; Cdh1; E-cad; Ecad; L-CAM; Um; UVO

            View more View less

            UniProt ID: (Mouse) P09803

            View more View less

            Entrez Gene ID: (Mouse) 12550

            View more View less

            Function(s)

            glycoprotein binding calcium ion binding protein binding beta-catenin binding protein phosphatase binding protein domain specific binding ankyrin binding GTPase activating protein binding gamma-catenin binding metal ion binding cell adhesion molecule binding cadherin

            Process(es)

            in utero embryonic development trophectodermal cell differentiation epithelial cell morphogenesis cell adhesion homophilic cell adhesion via plasma membrane adhesion molecules synapse assembly sensory perception of sound single organismal cell-cell adhesion calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules protein metabolic process negative regulation of cell-cell adhesion regulation of protein localization regulation of water loss via skin adherens junction organization positive regulation of transcription factor import into nucleus regulation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of transcription, DNA-templated regulation of epithelial cell proliferation negative regulation of epithelial cell proliferation protein homooligomerization intestinal epithelial cell development salivary gland cavitation regulation of branching involved in salivary gland morphogenesis bicellular tight junction assembly cellular response to amino acid stimulus cellular response to lithium ion cellular response to indole-3-methanol protein localization to plasma membrane establishment of protein localization to plasma membrane negative regulation of canonical Wnt signaling pathway cochlea development regulation of protein localization to cell surface regulation of neuron migration
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-flzw4:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0