Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: HeLa, SH-SY5Y, Mouse lung, Mouse thymus
Immunogen sequence: CPEEVQEPDS PSEEPLPSTS TLCPSPDSAQ PSSSTDPSIM EPTASSVPAP APTPQQAPPP PSLVPLEATD SLLELPHPLL QQTEDQFLSP TLA
E2F2 is a member of the E2Ffamily of transcription factors. The E2F family plays a crucialrole in the control of cell cycle and action of tumor suppressorproteins and is also a target of the transforming proteins of smallDNA tumor viruses. The E2F proteins contain several evolutionallyconserved domains found in most members of the family. Thesedomains include a DNA binding domain, a dimerization domain whichdetermines interaction with the differentiation regulatedtranscription factor proteins (DP), a transactivation domainenriched in acidic amino acids, and a tumor suppressor proteinassociation domain which is embedded within the transactivationdomain. This protein and another 2 members, E2F1 and E2F3, have anadditional cyclin binding domain. This protein binds specificallyto retinoblastoma protein pRB in a cell-cycle dependent manner, andit exhibits overall 46% amino acid identity to E2F1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2F-2; Transcription factor E2F2
Gene Aliases: E2F-2; E2F2
UniProt ID: (Human) Q14209, (Mouse) P56931
Entrez Gene ID: (Human) 1870, (Mouse) 242705
Molecular Function: RNA metabolism protein nucleic acid metabolism protein general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support