Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
11172-1-AP is a rabbit polyclonal antibody raised against the full-length human EAF2 protein.
Immunogen sequence: MNSAAGFSH LDRRERVLKL GESFEKQPRC AFHTVRYDFK PASIDTSSEG YLEVGEGEQV TITLPNIEGS TPPVTVFKGS KKPYLKECIL IINHDTGECR LEKLSSNITV KKTRVEGSSK IQYRKEQQQQ QMWNSARTPN LVKHSPSEDK MSPASPIDDI ERELKAEASL MDQMSSCDSS SDSKSSSSSS SEDSSSDSED EDCKSSTSDT GNCVSGHPTM TQYRIPDIDA SHNRFRDNSG LLMNTLRNDL QLSESGSDSD D (1-260 aa encoded by BC014209)
EAF1 and EAF2 can form a complex with ELL that targets the ternary elongation complex of Pol II, stimulating the rate of elongation. In addition, EAF1 and EAF2 are important for the stability of the NuA4 histone acetyltransferase complex, which transcriptionally activates certain genes by acetylation of Histones H4 and H2A. Both EAF1 and EAF2 are ubiquitously expressed members of the EAF family that colocalize with ELL to the the Cajal bodies and nuclear speckles. EAF1 contains a C-terminal region rich in aspartic acid, glutamic acid and serine residues. EAF2 is an androgen-response gene and can act as a potent apoptosis inducer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ehrlich S-II transcriptional activator factor; ELL-associated factor 2; testosterone regulated apoptosis inducer and tumor suppressor; Testosterone-regulated apoptosis inducer and tumor suppressor protein
Gene Aliases: AW048865; BM-040; BM040; EAF2; Festa; FESTA-L; FESTA-S; TRAITS; U19
UniProt ID: (Human) Q96CJ1, (Mouse) Q91ZD6, (Rat) Q811X5
Entrez Gene ID: (Human) 55840, (Mouse) 106389, (Rat) 266787
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support