Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EBF2 belongs to the conserved Olf/EBF family of helix-loop-helix transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collier, Olf and EBF 2; Early B-cell factor 2; EBF-2; MET-mesencephalon-olfactory TF1; MET-mesencephalon-olfactory transcription factor 1; metencephalon-mesencephalnon-olfactory transcription factor 1; Metencephalon-mesencephalon-olfactory transcription factor 1; O/E-3; OLF-1/EBF-LIKE 3; Transcription factor COE2
Gene Aliases: COE2; D14Ggc1e; EBF-2; EBF2; Mmot1; O/E-3; OE-3
UniProt ID: (Human) Q9HAK2, (Mouse) O08792
Entrez Gene ID: (Human) 64641, (Rat) 361060, (Mouse) 13592
Molecular Function: DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support