Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ECHDC2 belongs to the enoyl-CoA hydratase/isomerase family. The exact function of ECHDC2 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: enoyl CoA hydratase domain containing 2; enoyl Coenzyme A hydratase domain containing 2; Enoyl-CoA hydratase domain-containing protein 2, mitochondrial
Gene Aliases: 1300017C12Rik; 2610009M20Rik; D4Ertd765e; ECHDC2; RGD1308525
UniProt ID: (Human) Q86YB7, (Mouse) Q3TLP5
Entrez Gene ID: (Human) 55268, (Rat) 298381, (Mouse) 52430
Molecular Function: hydratase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support