Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MVSVPTTWCS VALALLVALH EGKGQAAATL EQPASSSHAQ GTHLRLRRCS CSSWLDKECV YFCHLDIIWV NTPEQTAPYG LGNPPRRRRR SLPRRCQCSS ARDPACATFC LRRPWTEAGA VPSRKSPADV FQTGKTGATT GELLQRLRDI STVKSLFAKR QQEAMREPRS THSRWRKR
Endothelins are endothelium-derived vasoconstrictor peptides. Endothelin 2, also known as ET-2, is a protein that in humans is encoded by the EDN2 gene. The secreted peptide (~2kD, 21 amino acids) is produced as a result of endothelin-2 precursor (~20 kD) processing and functions as a ligand for endothelin receptors which upon peptide binding modulates a wide array of cell signaling events. Diseases associated with endothelins include aortic disease and patent ductus arteriosus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EDN 2; EDN-2; Endothelin-2; ET-2; PP ET-2; PPET2; preproendothelin 2; Preproendothelin-2
Gene Aliases: EDN2; ET-2; ET2; PPET2
UniProt ID: (Human) P20800
Entrez Gene ID: (Human) 1907
Molecular Function:
peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support