Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 11
EF-1 (elongation factor-1) is a multi-protein complex that is responsible for the delivery of aminoacyl-tRNAs to the ribosome. EF-1 gamma (elongation factor- 1 gamma), also known as EEF1G or GIG35, is a 437 amino acid subunit of the EF-1 complex. Expressed in stomach, pancreas, brain, lung, kidney, intestine, liver and spleen, EF-1 gamma contains an N-terminal Glutathione transferase domain which is thought to be involved in anchoring the complex to various cellular components. Additionally, EF-1 gamma may play a key role in the assembly of multiprotein complexes containing aminoacyl-tRNA synthetases. Increased expression of EF-1 gamma is associated with pancreatic cancer, suggesting a possible role for EF-1 gamma in the oncogenic transformation process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eEF-1B gamma; EF-1-gamma; Elongation factor 1-gamma; pancreatic tumor-related protein; PRO1608; translation elongation factor eEF-1 gamma chain
Gene Aliases: 2610301D06Rik; AA407312; EEF1G; EF1G; GIG35; PRO1608
UniProt ID: (Human) P26641, (Rat) Q68FR6, (Mouse) Q9D8N0
Entrez Gene ID: (Human) 1937, (Rat) 293725, (Mouse) 67160
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support