Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
SELB is a translation factor necessary for the incorporation of selenocysteine into proteins. It probably replaces EF-Tu for the insertion of selenocysteine directed by the UGA codon. SelB binds GTP and GDP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: elongation factor for selenoprotein translation; Elongation factor sec; Eukaryotic elongation factor, selenocysteine-tRNA-specific; selenocysteine (Sec)-specific eukaryotic elongation factor; Selenocysteine-specific elongation factor
Gene Aliases: EEFSEC; EFSEC; SELB
UniProt ID: (Human) P57772
Entrez Gene ID: (Human) 60678
Molecular Function:
translation elongation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support