Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QQSTEKAVAA RDKLMDRHQD ISKAFTKTDQ SKTNYISICK MQEVLEECGC SLTEGELTHL LNSWGVSRHD NAINYLDFLR AVENSKSTGA QPKEKEESMP INFATLNPQE VVRKIQEVVE SSQLALSTAF SALDKEDT
This gene encodes a protein which directly binds the oncogene DJ-1 and androgen receptor to form a ternary complex in cells. This binding protein recruits histone-deacetylase complexes in order to repress transcription activity of androgen receptor. This protein may also play a role in spermatogenesis and fertilization. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAP-binding protein complex-interacting protein 1; DJ-1-binding protein; DJBP; EF-hand calcium-binding domain-containing protein 6
Gene Aliases: dJ185D5.1; DJBP; EFCAB6; HSCBCIP1; KIAA1672
UniProt ID: (Human) Q5THR3
Entrez Gene ID: (Human) 64800
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support